Learn More
MSC MCR Safety Safety Glasses, Scratch-Resistant, Polycarbonate, ANSI Z87.1, 1 ea
Catalog No. 502834682
Encompass
The MCR Safety Checklite CL1 safety glasses, model CL110, are engineered for comfort and durability in everyday use or as visitor specifications. Weighing under 23 grams, they feature rugged polycarbonate lenses that filter 99.9% of harmful UV rays, ensuring reliable eye protection. A smaller frame design provides a secure fit for a variety of face sizes.
  • Lightweight construction for long-term wear
  • Smaller frame sized to fit smaller faces
  • Excellent orbital seal and fit
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Features exclusive Duramass® scratch-resistant coating
  • Meets ANSI/ISEA Z87.1-2025 standard
  • Passes MIL-PRF-32432 standards
  • CAN/CSA Z94.3 compliant
Catalog No. 50-283-4682 Supplier MSC Supplier No. 06988166
The MCR Safety Checklite CL1 safety glasses, model CL110, are engineered for comfort and durability in everyday use or as visitor specifications. Weighing under 23 grams, they feature rugged polycarbonate lenses that filter 99.9% of harmful UV rays, ensuring reliable eye protection. A smaller frame design provides a secure fit for a variety of face sizes.
  • Lightweight construction for long-term wear
  • Smaller frame sized to fit smaller faces
  • Excellent orbital seal and fit
  • Rugged polycarbonate lens filters 99.9% of harmful UV rays
  • Features exclusive Duramass® scratch-resistant coating
  • Meets ANSI/ISEA Z87.1-2025 standard
  • Passes MIL-PRF-32432 standards
  • CAN/CSA Z94.3 compliant