Learn More
MSC KleenGuard Safety Glasses, Scratch-Resistant, Polycarbonate, Clear Mirror Lenses, 1 ea
Catalog No. 502834744
Encompass
These safety glasses feature scratch-resistant, polycarbonate, clear mirror lenses and a full-framed design. They are suitable for indoor/outdoor use with a wrap-around design, providing 99.9% UVA/UVB/UVC protection. The lightweight design includes a nosepiece with integrated ridges for comfort and slippage prevention.
  • Indoor/outdoor glasses style
  • Scratch-resistant lens coating
  • Full-framed design
  • Wrap-around frame design
  • Clear mirror polycarbonate lenses
  • Black frame and temples
  • Universal size
  • Non-adjustable temples
  • Provides 99.9% UVA/UVB/UVC protection
  • Nosepiece with integrated ridges
  • Includes neck cord
  • Meets ANSI Z87.1+ standards
  • Recyclable through RightCycle™ Program
Catalog No. 50-283-4744 Supplier MSC Supplier No. 59829010
These safety glasses feature scratch-resistant, polycarbonate, clear mirror lenses and a full-framed design. They are suitable for indoor/outdoor use with a wrap-around design, providing 99.9% UVA/UVB/UVC protection. The lightweight design includes a nosepiece with integrated ridges for comfort and slippage prevention.
  • Indoor/outdoor glasses style
  • Scratch-resistant lens coating
  • Full-framed design
  • Wrap-around frame design
  • Clear mirror polycarbonate lenses
  • Black frame and temples
  • Universal size
  • Non-adjustable temples
  • Provides 99.9% UVA/UVB/UVC protection
  • Nosepiece with integrated ridges
  • Includes neck cord
  • Meets ANSI Z87.1+ standards
  • Recyclable through RightCycle™ Program