Learn More
MSC SAFETY GLASSES
Catalog No. 502834778
Encompass
These safety glasses feature a lightweight, unisex design, offering wraparound coverage for enhanced eye protection. Constructed with durable polycarbonate, they provide a comfortable, contoured fit. The clear, uncoated lenses absorb 99.9% of UVA, UVB, and UVC rays, meeting ANSI Z87.1-2015 standards for impact protection.
  • Lightweight construction ensures wearer comfort
  • Unisex style suitable for all users
  • High wraparound coverage provides extended protection
  • Comfortable, contoured fit
  • Economical option for eye protection
  • U6 rated polycarbonate lenses absorb 99.9% of UV rays
  • Meets ANSI Z87.1-2015 impact-rated protector requirement
Catalog No. 50-283-4778 Supplier MSC Supplier No. 91187641
These safety glasses feature a lightweight, unisex design, offering wraparound coverage for enhanced eye protection. Constructed with durable polycarbonate, they provide a comfortable, contoured fit. The clear, uncoated lenses absorb 99.9% of UVA, UVB, and UVC rays, meeting ANSI Z87.1-2015 standards for impact protection.
  • Lightweight construction ensures wearer comfort
  • Unisex style suitable for all users
  • High wraparound coverage provides extended protection
  • Comfortable, contoured fit
  • Economical option for eye protection
  • U6 rated polycarbonate lenses absorb 99.9% of UV rays
  • Meets ANSI Z87.1-2015 impact-rated protector requirement