Learn More
MSC KleenGuard Safety Glasses, Scratch-Resistant, Polycarbonate, Smoke Mirror Lenses, Full-Framed
Catalog No. 502834911
Encompass
These V30 Nemesis Safety Glasses feature scratch-resistant, polycarbonate lenses with a smoke mirror color and a full-framed, black, wrap-around design. They meet ANSI Z87.1+ standards, offer 99.9% UVA/UVB/UVC protection, and include a nosepiece with integrated ridges to divert sweat for comfort.
  • Scratch-resistant polycarbonate lenses.
  • Provides 99.9% UVA/UVB/UVC protection.
  • Meets ANSI Z87.1+ standards.
  • Extremely flexible design for long-term comfort.
  • Nosepiece with integrated ridges helps divert sweat and prevent slippage.
  • Eligible for the EOL recycling program.
  • Includes neck cord.
Catalog No. 50-283-4911 Supplier MSC Supplier No. 59829002
These V30 Nemesis Safety Glasses feature scratch-resistant, polycarbonate lenses with a smoke mirror color and a full-framed, black, wrap-around design. They meet ANSI Z87.1+ standards, offer 99.9% UVA/UVB/UVC protection, and include a nosepiece with integrated ridges to divert sweat for comfort.
  • Scratch-resistant polycarbonate lenses.
  • Provides 99.9% UVA/UVB/UVC protection.
  • Meets ANSI Z87.1+ standards.
  • Extremely flexible design for long-term comfort.
  • Nosepiece with integrated ridges helps divert sweat and prevent slippage.
  • Eligible for the EOL recycling program.
  • Includes neck cord.