Learn More
MSC HexArmor Safety Glasses, Over-the-Glasses, Clear Lens, Anti-Fog, Scratch-Resistant, 1 ea
Catalog No. 502834583
Encompass
HexArmor LT250 safety glasses offer over-the-glasses (OTG) protection with permanent anti-fog and scratch-resistant TruShield® coatings. Designed to fit most prescription glasses, they ensure clear vision and premium optical clarity through LTX® Lens Technology.
  • OTG design fits over most prescription glasses
  • Hinged side arms fold flat for easy storage
  • Adaptive soft-grip side arms for comfort and secure fit
  • Contoured nose bridge enhances comfort
  • Unrestricted peripheral vision
  • Soft earpieces limit ear pressure
  • LTX® Lens Technology for premium optical clarity
  • Offers 100% UV protection
  • TruShield® coatings provide permanent anti-fog and scratch resistance, 4x thicker
  • Optical Class One lenses, 100% visually inspected
  • Compliant with ANSI Z87.1.2020, CE EN166, and CSA Z94.3
Catalog No. 50-283-4583 Supplier MSC Supplier No. 41683350
HexArmor LT250 safety glasses offer over-the-glasses (OTG) protection with permanent anti-fog and scratch-resistant TruShield® coatings. Designed to fit most prescription glasses, they ensure clear vision and premium optical clarity through LTX® Lens Technology.
  • OTG design fits over most prescription glasses
  • Hinged side arms fold flat for easy storage
  • Adaptive soft-grip side arms for comfort and secure fit
  • Contoured nose bridge enhances comfort
  • Unrestricted peripheral vision
  • Soft earpieces limit ear pressure
  • LTX® Lens Technology for premium optical clarity
  • Offers 100% UV protection
  • TruShield® coatings provide permanent anti-fog and scratch resistance, 4x thicker
  • Optical Class One lenses, 100% visually inspected
  • Compliant with ANSI Z87.1.2020, CE EN166, and CSA Z94.3