Learn More
MSC PRO-SAFE Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, ANSI Z87.1, 1 ea
Catalog No. 502834631
Encompass
These safety glasses feature a frameless design with polycarbonate lenses that offer anti-fog and scratch-resistant coatings, providing clear vision and protection for various applications.
  • Anti-fog and scratch-resistant lens coatings
  • Polycarbonate lens material
  • Frameless, wrap-around design
  • UV protection
  • Meets ANSI Z87.1+ safety standards
  • Universal fit
Catalog No. 50-283-4631 Supplier MSC Supplier No. 93885085
These safety glasses feature a frameless design with polycarbonate lenses that offer anti-fog and scratch-resistant coatings, providing clear vision and protection for various applications.
  • Anti-fog and scratch-resistant lens coatings
  • Polycarbonate lens material
  • Frameless, wrap-around design
  • UV protection
  • Meets ANSI Z87.1+ safety standards
  • Universal fit