Learn More
MSC KleenGuard Safety Glasses, Polycarbonate, Anti-Fog, Scratch-Resistant, ANSI Z87.1+
Catalog No. 502834902
Encompass
KleenGuard V30 Maverick Safety Glasses provide reliable eye protection with a durable, lightweight, and modern design. They feature polycarbonate lenses with 99.9% UVA/UVB/UVC protection and are recyclable through The RightCycle™ Program.
  • Equipped with KleenVision anti-fog coating.
  • Lenses are scratch-resistant.
  • Full-framed, wrap-around design.
  • Clear polycarbonate lenses.
  • Provides 99.9% UVA/UVB/UVC protection.
  • Meets ANSI Z87.1+ compliance.
  • Features a universal fit.
Catalog No. 50-283-4902 Supplier MSC Supplier No. 47775788
KleenGuard V30 Maverick Safety Glasses provide reliable eye protection with a durable, lightweight, and modern design. They feature polycarbonate lenses with 99.9% UVA/UVB/UVC protection and are recyclable through The RightCycle™ Program.
  • Equipped with KleenVision anti-fog coating.
  • Lenses are scratch-resistant.
  • Full-framed, wrap-around design.
  • Clear polycarbonate lenses.
  • Provides 99.9% UVA/UVB/UVC protection.
  • Meets ANSI Z87.1+ compliance.
  • Features a universal fit.