Learn More
Diversified Spaces™ Safety Glasses
SCI_ED
Catalog No. S42374 Shop All Diversified Spaces Products
Change view
Click to view available options
Lens Coating:
Uncoated
Catalog No. Lens Coating
S42374 Uncoated
1 options
This item is not returnable. View return policy
Catalog No. S42374 Supplier Diversified Spaces™ Supplier No. SG72000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protection against liquids, dust and smoke.
  • Durable planos
  • Adjustable temples
  • Universal-fit nose bridges
  • Flat-fold side shields
Certifications
MAS Certified Green
Compliance
SEFA
Warranty and Services
Limited Lifetime Warranty
Specifications
Lens Coating Uncoated
Lens Type Durable planos
Lens Tint Clear
Product Type Integrated Protective Eyewear
Material Polycarbonate, Nylon, PVC, Carbon Steel
Disposable Reusable
Lens Material PVC
Frame Material Plastic
UV Protection None
Frame Type Adjustable Temples
Autoclavable Not Autoclavable