Learn More
MSC MCR Safety Safety Glasses, Anti-Fog, Polycarbonate, Clear, ANSI Z87.1+
Supplier:  MSC 04782629
Encompass
The Klondike KD1 Series safety glasses offer comfort and protection for various environments. They feature a clear anti-fog polycarbonate lens, a black nylon frame, adjustable temples, and a soft TPR nose pad. These glasses meet ANSI Z87.1+ standards for eye protection.
  • Anti-fog coating improves visibility in hot and humid conditions
  • Polycarbonate lens material for durability
  • Clear lenses provide unobstructed vision
  • Full-framed, wrap-around design for comprehensive coverage
  • Black nylon frame with adjustable temples for a secure fit
  • Soft secure TPR nose pad for comfort
  • Meets ANSI Z87.1+ standards for eye protection
  • Offers UV protection
  • Full orbital seal with dual lens design
Catalog No. 50-283-4966