Learn More
MSC HexArmor Safety Glasses, Anti-Fog, Polycarbonate, Clear Lenses, Half-Framed, ANSI Z87.1.2020
Catalog No. 502834971
Encompass
These safety glasses are engineered for clear vision and protection in challenging environments. Featuring an anti-fog coating, they are constructed from durable polycarbonate with clear, half-framed lenses. Designed for universal fit, these glasses offer blue light blocking technology and comply with ANSI Z87.1.2020 standards, making them suitable for a wide range of industrial and laboratory applications.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Durable polycarbonate lens material.
  • Clear lenses for optimal visibility.
  • Half-framed design with wrap-around frame.
  • Gray and yellow frame and temple colors.
  • Non-adjustable and flexible temples for comfort.
  • Provides UV protection.
  • Features blue light blocking.
  • Meets ANSI Z87.1.2020 safety standards.
  • Frame made from thermoplastic rubber.
Catalog No. 50-283-4971 Supplier MSC Supplier No. 32388571
These safety glasses are engineered for clear vision and protection in challenging environments. Featuring an anti-fog coating, they are constructed from durable polycarbonate with clear, half-framed lenses. Designed for universal fit, these glasses offer blue light blocking technology and comply with ANSI Z87.1.2020 standards, making them suitable for a wide range of industrial and laboratory applications.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Durable polycarbonate lens material.
  • Clear lenses for optimal visibility.
  • Half-framed design with wrap-around frame.
  • Gray and yellow frame and temple colors.
  • Non-adjustable and flexible temples for comfort.
  • Provides UV protection.
  • Features blue light blocking.
  • Meets ANSI Z87.1.2020 safety standards.
  • Frame made from thermoplastic rubber.