Learn More
MSC 3M Protective Eyewear, Polycarbonate, Clear, Anti-Fog, ANSI Z87.1-2020
Catalog No. 502835054
Encompass
3M protective eyewear features anti-fog and scratch-resistant, clear polycarbonate lenses and a frameless, wraparound design. Equipped with a Cord Control System (CCS) for earplugs and an optional foam gasket, it ensures enhanced protection and comfort.
  • Cord Control System (CCS) keeps eyewear and earplugs attached and untangled.
  • Wraparound design provides extended eye protection and unobstructed viewing.
  • Contoured temples offer a snug fit.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Meets ANSI-ISEA Z87.1-2020 Impact-Rated Protector requirements.
  • Optional foam gasket limits dust exposure and adds cushioning.
  • Anti-fog and scratch-resistant coatings enhance visibility and durability.
Catalog No. 50-283-5054 Supplier MSC Supplier No. 87655601
3M protective eyewear features anti-fog and scratch-resistant, clear polycarbonate lenses and a frameless, wraparound design. Equipped with a Cord Control System (CCS) for earplugs and an optional foam gasket, it ensures enhanced protection and comfort.
  • Cord Control System (CCS) keeps eyewear and earplugs attached and untangled.
  • Wraparound design provides extended eye protection and unobstructed viewing.
  • Contoured temples offer a snug fit.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Meets ANSI-ISEA Z87.1-2020 Impact-Rated Protector requirements.
  • Optional foam gasket limits dust exposure and adds cushioning.
  • Anti-fog and scratch-resistant coatings enhance visibility and durability.