Learn More
MSC 3M Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, Frameless, ANSI Z87.1+, CSA Z94.3
Catalog No. 502835307
Encompass
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses in a frameless, wrap-around design. They meet ANSI Z87.1+ and CSA Z94.3 standards. They incorporate Pressure Diffusion Temple Technology, allowing temples to naturally adjust to individual head sizes for comfortable, durable, and lightweight eye protection.
  • Padded temple touchpoints and soft, adjustable nosepads offer added comfort for long-term wear.
  • Secure, snug fit keeps eyewear in place without slippage.
  • Motion-inspired, contoured piping in a bold, modern color palette for style.
  • Optional anti-fog coating helps keep lenses free of moisture in hot and humid conditions.
  • Polycarbonate lenses absorb 99.9% UVA and UVB down to 280 nm.
  • Weighs less than 1 oz., providing comfortable, durable, lightweight eye protection.
Catalog No. 50-283-5307 Supplier MSC Supplier No. 68918473
These safety glasses feature anti-fog and scratch-resistant clear polycarbonate lenses in a frameless, wrap-around design. They meet ANSI Z87.1+ and CSA Z94.3 standards. They incorporate Pressure Diffusion Temple Technology, allowing temples to naturally adjust to individual head sizes for comfortable, durable, and lightweight eye protection.
  • Padded temple touchpoints and soft, adjustable nosepads offer added comfort for long-term wear.
  • Secure, snug fit keeps eyewear in place without slippage.
  • Motion-inspired, contoured piping in a bold, modern color palette for style.
  • Optional anti-fog coating helps keep lenses free of moisture in hot and humid conditions.
  • Polycarbonate lenses absorb 99.9% UVA and UVB down to 280 nm.
  • Weighs less than 1 oz., providing comfortable, durable, lightweight eye protection.