Learn More
MSC 3M Safety Glasses, Scratch-Resistant, Polycarbonate, Clear Lenses, ANSI Z87.1-2015
Catalog No. 502835441
Encompass
These safety glasses are an economical eyewear option that combines style, fit, and lightweight comfort. Their wraparound design provides extended eye protection and unobstructed viewing.
  • Lightweight construction ensures wearer comfort.
  • Wraparound design provides extended eye protection and unobstructed viewing.
  • Unisex styling fits a variety of facial profiles.
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Meets the Impact-Rated Protector requirement of the ANSI Z87.1-2015 standard.
Catalog No. 50-283-5441 Supplier MSC Supplier No. 07417918
These safety glasses are an economical eyewear option that combines style, fit, and lightweight comfort. Their wraparound design provides extended eye protection and unobstructed viewing.
  • Lightweight construction ensures wearer comfort.
  • Wraparound design provides extended eye protection and unobstructed viewing.
  • Unisex styling fits a variety of facial profiles.
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays.
  • Meets the Impact-Rated Protector requirement of the ANSI Z87.1-2015 standard.