Learn More
MSC HexArmor Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, ANSI Z87.1.2020
Catalog No. 502835444
Encompass
These HexArmor safety glasses offer premium eye protection with a modern wraparound, frameless design. They feature LTX Lens Technology for exceptional optical clarity and a wide field of vision. Dual-action permanent anti-fog and scratch-resistant coatings, four times thicker than standard, ensure lasting performance. Designed for comfort and durability, they include a soft brow guard, nose piece, dual-injection soft-grip side arms, integrated eye ventilation, and 100% UV protection.
  • LTX lens technology for premium optical clarity.
  • Dual-injection soft-grip side arms for a slip-free, stable fit.
  • Innovative brow guard and soft nose piece for enhanced comfort and protection.
  • Compliant with ANSI Z87.1.2020, High Impact Rated Z87+, CSA Z94.3, and CE EN166 standards.
  • Polycarbonate lens material.
  • Clear lenses.
Catalog No. 50-283-5444 Supplier MSC Supplier No. 48942940
These HexArmor safety glasses offer premium eye protection with a modern wraparound, frameless design. They feature LTX Lens Technology for exceptional optical clarity and a wide field of vision. Dual-action permanent anti-fog and scratch-resistant coatings, four times thicker than standard, ensure lasting performance. Designed for comfort and durability, they include a soft brow guard, nose piece, dual-injection soft-grip side arms, integrated eye ventilation, and 100% UV protection.
  • LTX lens technology for premium optical clarity.
  • Dual-injection soft-grip side arms for a slip-free, stable fit.
  • Innovative brow guard and soft nose piece for enhanced comfort and protection.
  • Compliant with ANSI Z87.1.2020, High Impact Rated Z87+, CSA Z94.3, and CE EN166 standards.
  • Polycarbonate lens material.
  • Clear lenses.