Learn More
MSC 3M Safety Glasses, Solus 1000 Series, Clear Polycarbonate, Anti-Fog/Scratch-Resistant
Catalog No. 502835486
Encompass
3M Solus 1000 Series Safety Glasses offer robust impact protection with a modern, sporty design. These glasses feature a Scotchgard Anti-Fog coating, providing superior clarity and scratch resistance, making them ideal for challenging work environments.
  • Scotchgard Protector Anti-Fog Coating resists fogging and provides scratch resistance through multiple washings.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays for comprehensive UV protection.
  • Padded temples and a soft nose bridge ensure enhanced comfort during prolonged wear.
  • Configurable design allows for detachable gasket, strap, or temple options to suit various needs.
  • Meets ANSI-ISEA Z87.1-2020 standard for impact and anti-fog performance.
Catalog No. 50-283-5486 Supplier MSC Supplier No. 66949587
3M Solus 1000 Series Safety Glasses offer robust impact protection with a modern, sporty design. These glasses feature a Scotchgard Anti-Fog coating, providing superior clarity and scratch resistance, making them ideal for challenging work environments.
  • Scotchgard Protector Anti-Fog Coating resists fogging and provides scratch resistance through multiple washings.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays for comprehensive UV protection.
  • Padded temples and a soft nose bridge ensure enhanced comfort during prolonged wear.
  • Configurable design allows for detachable gasket, strap, or temple options to suit various needs.
  • Meets ANSI-ISEA Z87.1-2020 standard for impact and anti-fog performance.