Learn More
MSC PRO-SAFE Safety Glasses, Scratch-Resistant, Polycarbonate, Clear Lenses, Frameless
Catalog No. 502834998
Encompass
These safety glasses are designed to protect against dangerous particles, featuring scratch-resistant polycarbonate clear lenses in a frameless, wrap-around style. They offer a universal fit and meet ANSI Z87.1+ standards for protective eyewear.
  • Scratch-resistant lenses
  • Polycarbonate lens material
  • Clear lenses
  • Frameless design
  • Wrap around style for enhanced protection
  • Universal size for a comfortable fit
  • Meets ANSI Z87.1+ safety standards
  • Provides UV protection
  • Flexible temples
Catalog No. 50-283-4998 Supplier MSC Supplier No. 93885069
These safety glasses are designed to protect against dangerous particles, featuring scratch-resistant polycarbonate clear lenses in a frameless, wrap-around style. They offer a universal fit and meet ANSI Z87.1+ standards for protective eyewear.
  • Scratch-resistant lenses
  • Polycarbonate lens material
  • Clear lenses
  • Frameless design
  • Wrap around style for enhanced protection
  • Universal size for a comfortable fit
  • Meets ANSI Z87.1+ safety standards
  • Provides UV protection
  • Flexible temples