Learn More
MSC MCR Safety Safety Glasses, Polycarbonate, Clear Lens, Scratch-Resistant, ANSI Z87.1+
Catalog No. 502835040
Encompass
MCR Safety Klondike KD1 series safety glasses feature clear polycarbonate lenses with a scratch-resistant coating, designed for various applications. They offer a full-framed, wrap-around style with adjustable spatula temples and a soft TPR nose pad for comfort and security.
  • Full orbital seal with dual-lens design
  • Matte black frame finish
  • Soft, secure TPR nose pad
  • Extendable spatula temples
  • Adjustable temples
  • UV protection
  • ANSI Z87.1+ compliant
Catalog No. 50-283-5040 Supplier MSC Supplier No. 04782611
MCR Safety Klondike KD1 series safety glasses feature clear polycarbonate lenses with a scratch-resistant coating, designed for various applications. They offer a full-framed, wrap-around style with adjustable spatula temples and a soft TPR nose pad for comfort and security.
  • Full orbital seal with dual-lens design
  • Matte black frame finish
  • Soft, secure TPR nose pad
  • Extendable spatula temples
  • Adjustable temples
  • UV protection
  • ANSI Z87.1+ compliant