Learn More
MSC PRO-SAFE Safety Glasses, Scratch-Resistant, Polycarbonate, Frameless, ANSI Z87.1+
Catalog No. 502835099
Encompass
PRO-SAFE safety glasses are designed for high-speed applications and general eye protection in various work environments. These durable and comfortable safety glasses feature scratch-resistant polycarbonate lenses and a lightweight, frameless, wraparound design, making them suitable for everyday use and for visitors in industrial settings.
  • Scratch-resistant polycarbonate lenses
  • Frameless, wraparound design for comprehensive coverage
  • Non-adjustable, flexible temples for comfortable wear
  • Lightweight construction ensures comfort during extended use
  • Universal size suitable for a wide range of users
  • Provides UV protection
  • Complies with ANSI Z87.1+ safety standards
Catalog No. 50-283-5099 Supplier MSC Supplier No. 93884351
PRO-SAFE safety glasses are designed for high-speed applications and general eye protection in various work environments. These durable and comfortable safety glasses feature scratch-resistant polycarbonate lenses and a lightweight, frameless, wraparound design, making them suitable for everyday use and for visitors in industrial settings.
  • Scratch-resistant polycarbonate lenses
  • Frameless, wraparound design for comprehensive coverage
  • Non-adjustable, flexible temples for comfortable wear
  • Lightweight construction ensures comfort during extended use
  • Universal size suitable for a wide range of users
  • Provides UV protection
  • Complies with ANSI Z87.1+ safety standards