Learn More
MSC HexArmor Safety Glasses, VS300, TruShield2F Anti-Fog, ANSI Z87.1.2020 Compliant
Catalog No. 502835136
Encompass
The HexArmor VS300 anti-fog safety glasses provide optical clarity and full coverage protection with a comfortable fit. They feature dual-action permanent anti-fog and scratch-resistant TruShield2F coatings, optimized for hot to cold temperature transitions, which are significantly thicker and more durable than competitors' coatings. Each lens is 100% visually inspected for optical class one clarity.
  • Soft ear pieces for stable, low-pressure fit.
  • Brow guard protects from flying particles, dirt, and dust.
  • Flexible side arms with seven inclination levels.
  • Triple-injection hard and soft components for durability and comfort.
  • Soft flex nose loops for even weight distribution.
  • LTX Lens Technology for premium optical clarity.
  • 100% UV protection and metal free design.
  • Meets ANSI Z87.1.2020, High Impact Z87+, CE EN166, and CSA Z94.3 compliance.
Catalog No. 50-283-5136 Supplier MSC Supplier No. 32169401
The HexArmor VS300 anti-fog safety glasses provide optical clarity and full coverage protection with a comfortable fit. They feature dual-action permanent anti-fog and scratch-resistant TruShield2F coatings, optimized for hot to cold temperature transitions, which are significantly thicker and more durable than competitors' coatings. Each lens is 100% visually inspected for optical class one clarity.
  • Soft ear pieces for stable, low-pressure fit.
  • Brow guard protects from flying particles, dirt, and dust.
  • Flexible side arms with seven inclination levels.
  • Triple-injection hard and soft components for durability and comfort.
  • Soft flex nose loops for even weight distribution.
  • LTX Lens Technology for premium optical clarity.
  • 100% UV protection and metal free design.
  • Meets ANSI Z87.1.2020, High Impact Z87+, CE EN166, and CSA Z94.3 compliance.