Learn More
MSC Radians Safety Eyewear, Foam Lined, Clear Polycarbonate Lens, Anti-Fog, ANSI Z87.1+
Catalog No. 502835138
Encompass
Radians Mirage™ Foam Safety Eyewear is a foam-lined safety glass designed to block debris, airborne particles, and eye irritants. It offers sleek, light, and comfortable eye protection that utilizes foam-lined vents to filter air for maximum protection.
  • Closed-cell foam-lined channels block debris and eye irritants.
  • Full wraparound frame allows for unobstructed peripheral vision.
  • Tiny foam-lined pockets/vents provide extra fog control.
  • Polycarbonate lens provides impact protection.
  • Meets ANSI Z87.1+ standards.
Catalog No. 50-283-5138 Supplier MSC Supplier No. 70086913
Radians Mirage™ Foam Safety Eyewear is a foam-lined safety glass designed to block debris, airborne particles, and eye irritants. It offers sleek, light, and comfortable eye protection that utilizes foam-lined vents to filter air for maximum protection.
  • Closed-cell foam-lined channels block debris and eye irritants.
  • Full wraparound frame allows for unobstructed peripheral vision.
  • Tiny foam-lined pockets/vents provide extra fog control.
  • Polycarbonate lens provides impact protection.
  • Meets ANSI Z87.1+ standards.