Learn More
MSC HexArmor Safety Glasses, Variomatic, Polycarbonate, Anti-Fog, ANSI Z87.1.2020
Catalog No. 502835168
Encompass
HexArmor VS250 Safety Glasses feature Variomatic, anti-fog, and scratch-resistant polycarbonate lenses in a half-framed design. These lenses appear clear and turn blue in sunlight. They provide 100% UV protection and are compliant with ANSI Z87.1.2020, offering premium optical clarity and adaptability for various face shapes.
  • Lightweight (23 grams)
  • Soft, non-slip, adaptive side arms for optimum comfort
  • Contoured lens delivers a wide field of vision
  • Adjustable nose piece provides a comfortable and customized fit
  • Lens technology ensures premium optical clarity
  • 100% UV protection
  • Dual-action anti-fog and scratch-resistant coatings are 4x thicker than standard
  • Adaptable to most face shapes for high adaptability, durability, and comfort
Catalog No. 50-283-5168 Supplier MSC Supplier No. 46988309
HexArmor VS250 Safety Glasses feature Variomatic, anti-fog, and scratch-resistant polycarbonate lenses in a half-framed design. These lenses appear clear and turn blue in sunlight. They provide 100% UV protection and are compliant with ANSI Z87.1.2020, offering premium optical clarity and adaptability for various face shapes.
  • Lightweight (23 grams)
  • Soft, non-slip, adaptive side arms for optimum comfort
  • Contoured lens delivers a wide field of vision
  • Adjustable nose piece provides a comfortable and customized fit
  • Lens technology ensures premium optical clarity
  • 100% UV protection
  • Dual-action anti-fog and scratch-resistant coatings are 4x thicker than standard
  • Adaptable to most face shapes for high adaptability, durability, and comfort