Learn More
MSC MCR Safety Safety Glasses, Polycarbonate, Clear Lens, ANSI Z87.1+
Catalog No. 502835170
Encompass
The BearKat BK1 safety glasses offer a blend of safety, comfort, and value. They feature a one-piece wraparound lens that provides an unobstructed view and a secure fit for maximum protection. Flexible temples and non-slip sleeves enhance comfort, while notched ends accommodate a clip-on breakaway cord.
  • Strong yet lightweight design
  • Single lens design with integral side shields
  • Non-slip soft temple material
  • Exclusive Duramass scratch resistant coating
  • Rugged polycarbonate lens
  • Filters 99.9% of harmful UV rays
  • Meets ANSI Z87+ and CSA Z94.3 standards
Catalog No. 50-283-5170 Supplier MSC Supplier No. 76652007
The BearKat BK1 safety glasses offer a blend of safety, comfort, and value. They feature a one-piece wraparound lens that provides an unobstructed view and a secure fit for maximum protection. Flexible temples and non-slip sleeves enhance comfort, while notched ends accommodate a clip-on breakaway cord.
  • Strong yet lightweight design
  • Single lens design with integral side shields
  • Non-slip soft temple material
  • Exclusive Duramass scratch resistant coating
  • Rugged polycarbonate lens
  • Filters 99.9% of harmful UV rays
  • Meets ANSI Z87+ and CSA Z94.3 standards