Learn More
MSC PRO-SAFE Safety Glasses, Anti-Fog, Scratch-Resistant, Polycarbonate, Half-Framed, ANSI Z87.1+
Catalog No. 502835322
Encompass
These safety glasses are designed with anti-fog and scratch-resistant polycarbonate clear lenses in a half-framed style. They provide reliable eye protection and meet ANSI Z87.1+ compliance standards.
  • Anti-fog coating improves visibility in hot and humid conditions
  • Scratch-resistant lenses for enhanced durability
  • Polycarbonate clear lenses offer broad visibility
  • Half-framed design with black plastic and rubber frame
  • Non-adjustable temples for a secure fit
  • Provides UV protection
  • Complies with ANSI Z87.1+ safety standards
  • Universal size with a wrap-around style
Catalog No. 50-283-5322 Supplier MSC Supplier No. 43372440
These safety glasses are designed with anti-fog and scratch-resistant polycarbonate clear lenses in a half-framed style. They provide reliable eye protection and meet ANSI Z87.1+ compliance standards.
  • Anti-fog coating improves visibility in hot and humid conditions
  • Scratch-resistant lenses for enhanced durability
  • Polycarbonate clear lenses offer broad visibility
  • Half-framed design with black plastic and rubber frame
  • Non-adjustable temples for a secure fit
  • Provides UV protection
  • Complies with ANSI Z87.1+ safety standards
  • Universal size with a wrap-around style