Learn More
MSC 3M Safety Glasses, Anti-Fog, Scratch-Resistant, Clear Lens, Half-Framed
Catalog No. 502835064
Encompass
The Solus 1000 Series Safety Glasses Kit provides reliable eye protection for challenging work environments. Featuring a durable anti-fog and scratch-resistant coating, these glasses ensure clearer vision and enhanced performance. This kit is designed for comfort and extended use in various industrial and laboratory settings.
  • Advanced anti-fog coating resists fogging longer and helps provide scratch resistance.
  • Kit includes removable temples, an optional elastic strap, and a removable gasket.
  • Contemporary styling with padded temples and a soft nose bridge for enhanced comfort.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Meets ANSI-ISEA Z87.1-2020 Standard and CSA Z94.3 for impact protection and anti-fog performance.
  • Coating retains effectiveness for at least 25 washings.
Catalog No. 50-283-5064 Supplier MSC Supplier No. 66949637
The Solus 1000 Series Safety Glasses Kit provides reliable eye protection for challenging work environments. Featuring a durable anti-fog and scratch-resistant coating, these glasses ensure clearer vision and enhanced performance. This kit is designed for comfort and extended use in various industrial and laboratory settings.
  • Advanced anti-fog coating resists fogging longer and helps provide scratch resistance.
  • Kit includes removable temples, an optional elastic strap, and a removable gasket.
  • Contemporary styling with padded temples and a soft nose bridge for enhanced comfort.
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Meets ANSI-ISEA Z87.1-2020 Standard and CSA Z94.3 for impact protection and anti-fog performance.
  • Coating retains effectiveness for at least 25 washings.