Learn More
MSC 3M Safety Glasses, Polycarbonate, Anti-Fog, Scratch-Resistant, ANSI Z87.1-2015
Catalog No. 502835167
Encompass
These safety glasses from the SecureFit 400 Series feature clear polycarbonate lenses with anti-fog and scratch-resistant coatings, designed to offer reliable eye protection in various work environments. They meet both ANSI Z87.1-2015 and CSA Z94.3 standards, ensuring high levels of safety compliance. The frameless, wrap-around design provides a wide field of vision, and they incorporate Pressure Diffusion Temple Technology for enhanced comfort.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Lenses are scratch-resistant.
  • Clear polycarbonate lenses offer durability and clarity.
  • Frameless, wrap-around design for a wide field of vision.
  • Provides UV protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 safety standards.
  • Features Pressure Diffusion Temple Technology for enhanced comfort.
Catalog No. 50-283-5167 Supplier MSC Supplier No. 46722260
These safety glasses from the SecureFit 400 Series feature clear polycarbonate lenses with anti-fog and scratch-resistant coatings, designed to offer reliable eye protection in various work environments. They meet both ANSI Z87.1-2015 and CSA Z94.3 standards, ensuring high levels of safety compliance. The frameless, wrap-around design provides a wide field of vision, and they incorporate Pressure Diffusion Temple Technology for enhanced comfort.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Lenses are scratch-resistant.
  • Clear polycarbonate lenses offer durability and clarity.
  • Frameless, wrap-around design for a wide field of vision.
  • Provides UV protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 safety standards.
  • Features Pressure Diffusion Temple Technology for enhanced comfort.