Learn More
MSC PYRAMEX Safety Glasses, Polycarbonate, Clear Lens, Frameless, ANSI Z87.1
Catalog No. 502835223
Encompass
The Intruder safety glasses offer ultimate protection and comfort with a lightweight, stylish design. They provide superior impact and debris protection, and feature an integrated nose piece for a secure fit. The scratch-resistant lenses ensure clarity in any environment. The frameless design offers unparalleled comfort, making them ideal for long hours. A panoramic polycarbonate, wraparound lens provides an extended field of vision and UV protection. Some styles are treated with anti-fog properties for fog-free clarity.
  • Frameless, lightweight design
  • Panoramic polycarbonate, wraparound lens
  • Integrated nose piece for a secure fit
  • Scratch-resistant lenses
  • UV protection
  • Meets ANSI/ISEA Z87.1-2020 and CAN/CSA Z94.3-20 standards
Catalog No. 50-283-5223 Supplier MSC Supplier No. 68912609
The Intruder safety glasses offer ultimate protection and comfort with a lightweight, stylish design. They provide superior impact and debris protection, and feature an integrated nose piece for a secure fit. The scratch-resistant lenses ensure clarity in any environment. The frameless design offers unparalleled comfort, making them ideal for long hours. A panoramic polycarbonate, wraparound lens provides an extended field of vision and UV protection. Some styles are treated with anti-fog properties for fog-free clarity.
  • Frameless, lightweight design
  • Panoramic polycarbonate, wraparound lens
  • Integrated nose piece for a secure fit
  • Scratch-resistant lenses
  • UV protection
  • Meets ANSI/ISEA Z87.1-2020 and CAN/CSA Z94.3-20 standards