Learn More
S-Curve™ Safety Glasses Dispenser with Tray
Catalog No. 01182480 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
PETG
Catalog No. Material
01-182-480 PETG
1 options
Catalog No. 01-182-480 Supplier S-Curve™ Supplier No. BD614PETG
Add to Cart
Add to Cart
This clear Safety Glass Dispenser is perfect for bulk safety glasses. It can be wall mounted and the PETG material is chemical and impact resistant.
  • BD-614-PETG Bulk Safety Glasses Dispenser
  • 1 compartment dispenser with front access
  • 3/16 in. clear PETG material - SAFE with IPA
  • Mounts to wall or free-standing
  • Flame polished edges for a smooth finish
  • Product Size: 8W x 14H x 6.5D in.
  • 1 per case
Specifications
Product Type Dispenser
Material PETG
Disposable Reusable