Learn More
Glass Expansion Safety Gloves, Large
SDP
Catalog No. 502702946
Encompass_Preferred
Safety Gloves, Large. These safety gloves are provided with the D-Torch for use while handling the D-Torch. Always wear safety gloves and safety glasses when handling ICP torches because they contain glass. Other safety glove sizes available are small (P/N SAFEGLOVE-sml) and medium (P/N SAFEGLOVE-med). Safety glasses are also available (P/N SAFEGLASS). Safety first!Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2946 Supplier Glass Expansion Supplier No. SAFEGLOVELGE
Safety Gloves, Large. These safety gloves are provided with the D-Torch for use while handling the D-Torch. Always wear safety gloves and safety glasses when handling ICP torches because they contain glass. Other safety glove sizes available are small (P/N SAFEGLOVE-sml) and medium (P/N SAFEGLOVE-med). Safety glasses are also available (P/N SAFEGLASS). Safety first!Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001