Learn More
Glass Expansion Safety Gloves, Medium
SDP
Catalog No. 502702945
Encompass_Preferred
Safety Gloves, Medium. These safety gloves are provided with the D-Torch for use while handling the D-Torch. Always wear safety gloves and safety glasses when handling ICP torches because they contain glass. Other safety glove sizes available are small (P/N SAFEGLOVE-sml) and large (P/N SAFEGLOVE-lge). Safety glasses are also available (P/N SAFEGLASS). Safety first!Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2945 Supplier Glass Expansion Supplier No. SAFEGLOVEMED
Safety Gloves, Medium. These safety gloves are provided with the D-Torch for use while handling the D-Torch. Always wear safety gloves and safety glasses when handling ICP torches because they contain glass. Other safety glove sizes available are small (P/N SAFEGLOVE-sml) and large (P/N SAFEGLOVE-lge). Safety glasses are also available (P/N SAFEGLASS). Safety first!Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001