Learn More
MSC SAFETY GLS
Catalog No. 5000792749
Encompass
These safety glasses are designed for eye protection, featuring scratch-resistant, polycarbonate gray lenses in a frameless design with a black frame. They are suitable for various industrial and laboratory environments, meeting CSA Z94.3 and ANSI Z87.1-2015 standards.
  • Scratch-resistant lenses for enhanced durability
  • Polycarbonate lens material provides impact resistance
  • Gray lenses offer UV protection and reduce glare
  • Frameless design with black frame for a modern look
  • Non-adjustable temples with 3M Pressure Diffusion Temple Technology for a secure and comfortable fit
  • Meets CSA Z94.3 and ANSI Z87.1-2015 safety standards
Catalog No. 50-007-92749 Supplier MSC Supplier No. 96198262
These safety glasses are designed for eye protection, featuring scratch-resistant, polycarbonate gray lenses in a frameless design with a black frame. They are suitable for various industrial and laboratory environments, meeting CSA Z94.3 and ANSI Z87.1-2015 standards.
  • Scratch-resistant lenses for enhanced durability
  • Polycarbonate lens material provides impact resistance
  • Gray lenses offer UV protection and reduce glare
  • Frameless design with black frame for a modern look
  • Non-adjustable temples with 3M Pressure Diffusion Temple Technology for a secure and comfortable fit
  • Meets CSA Z94.3 and ANSI Z87.1-2015 safety standards