Learn More
MSC SAFETY GLS
Catalog No. 5000688469
Encompass
These safety glasses feature anti-fog and scratch-resistant polycarbonate lenses, designed to provide clear vision and protection in various indoor and outdoor working conditions.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Scratch-resistant lenses enhance durability.
  • Polycarbonate lens material for impact resistance.
  • Clear mirror lenses suitable for indoor/outdoor use.
  • Full-framed design with black rubber frame.
  • Integrated side shields for enhanced protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Universal size with non-adjustable black temples.
  • Provides UV protection.
Catalog No. 50-006-88469 Supplier MSC Supplier No. 96198189
These safety glasses feature anti-fog and scratch-resistant polycarbonate lenses, designed to provide clear vision and protection in various indoor and outdoor working conditions.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Scratch-resistant lenses enhance durability.
  • Polycarbonate lens material for impact resistance.
  • Clear mirror lenses suitable for indoor/outdoor use.
  • Full-framed design with black rubber frame.
  • Integrated side shields for enhanced protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Universal size with non-adjustable black temples.
  • Provides UV protection.