Learn More
MSC SAFETY GLS
Catalog No. 5000676376
Encompass
These safety glasses feature anti-fog and scratch-resistant polycarbonate lenses, designed for optimal clarity and durability. They are constructed with a full-framed design and comply with the ANSI Z87.1-2015 standard.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Blue mirror lens color
  • Full-framed, wrap-around design with black frame
  • Non-adjustable black temples
  • UV protection
  • Polarized lenses
  • Complies with ANSI Z87.1-2015 standard
Catalog No. 50-006-76376 Supplier MSC Supplier No. 96198221
These safety glasses feature anti-fog and scratch-resistant polycarbonate lenses, designed for optimal clarity and durability. They are constructed with a full-framed design and comply with the ANSI Z87.1-2015 standard.
  • Anti-fog and scratch-resistant polycarbonate lenses
  • Blue mirror lens color
  • Full-framed, wrap-around design with black frame
  • Non-adjustable black temples
  • UV protection
  • Polarized lenses
  • Complies with ANSI Z87.1-2015 standard