Learn More
Alkali Scientific Safety Goggles with Clear Anti Fog Lens, can fit over regular optical eyewear, 1ea
SDP
Catalog No. 5000663302
Encompass_Preferred
For users who require safety protection over prescription glasses, the Law OTG (Over the Glass) safety eyewear provides a practical and comfortable solution. The over d frame and large lens design fit easily over most prescription eyewear while maintaining clear visibility. Non slip TPR temples enhance comfort and provide a secure grip during extended wear. The wraparound style improves angular coverage and peripheral vision, offering better protection in laboratory and industrial environments. A five position ratchet temple mechanism allows the user to adjust the lens angle for a customized fit. The dielectric design contains no metal parts, making the eyewear suitable for environments where electrical safety is required. Durable polycarbonate lenses are equipped with exclusive Duramass scratch resistant coating and filter 99.9% of harmful ultraviolet rays.
Catalog No. 50-006-63302 Supplier Alkali Scientific Supplier No. G750
For users who require safety protection over prescription glasses, the Law OTG (Over the Glass) safety eyewear provides a practical and comfortable solution. The over d frame and large lens design fit easily over most prescription eyewear while maintaining clear visibility. Non slip TPR temples enhance comfort and provide a secure grip during extended wear. The wraparound style improves angular coverage and peripheral vision, offering better protection in laboratory and industrial environments. A five position ratchet temple mechanism allows the user to adjust the lens angle for a customized fit. The dielectric design contains no metal parts, making the eyewear suitable for environments where electrical safety is required. Durable polycarbonate lenses are equipped with exclusive Duramass scratch resistant coating and filter 99.9% of harmful ultraviolet rays.