Learn More
Med Vet International Safety Goggles, Vented
Catalog No. 502830768
Encompass_Preferred
Soft PVC vinyl frame with polycarbonate lens fits face comfortably with an elastic headband. Will fit over regular eyeglasses. Ventilation prevents fogging. They provide protection from particulates while offering a wider view of the work area than traditional impact goggles.
Catalog No. 50-283-0768 Supplier Med Vet International Supplier No. 972001
Soft PVC vinyl frame with polycarbonate lens fits face comfortably with an elastic headband. Will fit over regular eyeglasses. Ventilation prevents fogging. They provide protection from particulates while offering a wider view of the work area than traditional impact goggles.