Learn More
MSC SAFETY HELMET
Catalog No. 5000685032
Encompass
This Milwaukee Tool hard hat is designed for head protection in demanding environments, offering impact resistance and climbing-style features for enhanced safety.
  • Climbing style for specialized applications
  • Impact-resistant design for enhanced head protection
  • Constructed from durable polyethylene material
  • ANSI Type II compliant
  • Class E rating for electrical protection
  • Non-vented design
  • Non-slotted for secure attachment of accessories
  • Universal size for broad compatibility
  • Features a 4-point suspension system for comfort and stability
  • Adjustable for a customized fit
  • Blue color
Catalog No. 50-006-85032 Supplier MSC Supplier No. 10880771
This Milwaukee Tool hard hat is designed for head protection in demanding environments, offering impact resistance and climbing-style features for enhanced safety.
  • Climbing style for specialized applications
  • Impact-resistant design for enhanced head protection
  • Constructed from durable polyethylene material
  • ANSI Type II compliant
  • Class E rating for electrical protection
  • Non-vented design
  • Non-slotted for secure attachment of accessories
  • Universal size for broad compatibility
  • Features a 4-point suspension system for comfort and stability
  • Adjustable for a customized fit
  • Blue color