Learn More
Grainger Safety Interlock Switch Plastic 2NC
Catalog No. 502752818
Encompass_Preferred
The Telemecanique XCSLE-14L755 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. The operating temperature range is -25°C to 70°C. This interlock switch is designed for use in safety applications up to PLe/Cat.4 per ISO 13849-1. It features direct opening action and is tamper resistant. The switch provides position interlock detection for moving guards on machinery.
Catalog No. 50-275-2818 Supplier Grainger Supplier No. 14L755
The Telemecanique XCSLE-14L755 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. The operating temperature range is -25°C to 70°C. This interlock switch is designed for use in safety applications up to PLe/Cat.4 per ISO 13849-1. It features direct opening action and is tamper resistant. The switch provides position interlock detection for moving guards on machinery.