Learn More
Grainger Safety Interlock Switch Zamak 1NO/2NC
Catalog No. 502752658
Encompass_Preferred
The Telemecanique XCSLF-14L750 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. The operating temperature range is -25°C to 70°C. This interlock switch is designed for use in safety applications up to PLe/Cat.4 per ISO 13849-1. It features direct opening action and comes with a 300mm cable. The XCSLF series offers various actuator lengths from 5mm to 110mm.
Catalog No. 50-275-2658 Supplier Grainger Supplier No. 14L750
The Telemecanique XCSLF-14L750 is a safety interlock switch with a 14mm actuator. It has a plastic housing rated IP67 with 2 NC contacts. The operating temperature range is -25°C to 70°C. This interlock switch is designed for use in safety applications up to PLe/Cat.4 per ISO 13849-1. It features direct opening action and comes with a 300mm cable. The XCSLF series offers various actuator lengths from 5mm to 110mm.