Learn More
Grainger Safety Interlock Switch Zamak 1NO/2NC
Catalog No. 502752797
Encompass_Preferred
The Grainger website features mechanically actuated safety interlock switches. These switches are designed to help protect personnel by shutting down machinery when guards are opened. They feature direct opening contacts and come in various actuator styles including plunger, roller plunger, and tongue actuators. The switches have an epoxy sealed housing for protection in washdown environments. They are available in multiple contact configurations and can be wired for normally open or normally closed operation. The switches meet OSHA and ANSI machine safety requirements.
Catalog No. 50-275-2797 Supplier Grainger Supplier No. 14L749
The Grainger website features mechanically actuated safety interlock switches. These switches are designed to help protect personnel by shutting down machinery when guards are opened. They feature direct opening contacts and come in various actuator styles including plunger, roller plunger, and tongue actuators. The switches have an epoxy sealed housing for protection in washdown environments. They are available in multiple contact configurations and can be wired for normally open or normally closed operation. The switches meet OSHA and ANSI machine safety requirements.