Learn More
Grainger Safety Interlock Switch Zamak 2NO/1NC
Catalog No. 502749820
Encompass_Preferred
The Grainger door hinge safety interlock switches are designed to stop machine operation when a guard door is opened. They feature a compact design that mounts directly to the door hinge. The switches have a rugged die-cast housing and are available in several configurations including normally open, normally closed, and 2NC/2NO. These interlock switches are UL listed and CSA certified. They can handle up to 10A at 480VAC. The switches provide a cost-effective method of implementing safety interlocks.
Catalog No. 50-274-9820 Supplier Grainger Supplier No. 14L745
The Grainger door hinge safety interlock switches are designed to stop machine operation when a guard door is opened. They feature a compact design that mounts directly to the door hinge. The switches have a rugged die-cast housing and are available in several configurations including normally open, normally closed, and 2NC/2NO. These interlock switches are UL listed and CSA certified. They can handle up to 10A at 480VAC. The switches provide a cost-effective method of implementing safety interlocks.