Learn More
Grainger Safety Interlock Switch Zamak 1NO/2NC
Catalog No. 502751656
Encompass_Preferred
The TESXCSB703 is a safety interlock switch made by Schneider Electric. It has a zinc alloy body and 1 normally open and 2 normally closed contacts. This interlock switch is designed to fit 22mm diameter actuators. It has gold plated double wipe contacts rated for 10A at 250VAC. The operating temperature range is -25°C to 85°C. This interlock switch provides position interlocking for moving guards on machinery. It features a rugged metal construction and ensures the power is switched off when a guard is opened.
Catalog No. 50-275-1656 Supplier Grainger Supplier No. 14L751
The TESXCSB703 is a safety interlock switch made by Schneider Electric. It has a zinc alloy body and 1 normally open and 2 normally closed contacts. This interlock switch is designed to fit 22mm diameter actuators. It has gold plated double wipe contacts rated for 10A at 250VAC. The operating temperature range is -25°C to 85°C. This interlock switch provides position interlocking for moving guards on machinery. It features a rugged metal construction and ensures the power is switched off when a guard is opened.