Learn More
Grainger Safety Interlock Switch Zamak 1NO/2NC
Catalog No. 502751659
Encompass_Preferred
The Telemecanique XCSLF-14L752 is a safety interlock switch with a metal head and plastic body. It has a 14mm actuator with a maximum switching capacity of 6A at 250V AC. This interlock switch has gold plated contacts and is IP67 rated for protection against dust and temporary immersion. It is designed for use on safety guards and access panels to prevent operation when open. This interlock switch provides reliable position detection for industrial safety applications.
Catalog No. 50-275-1659 Supplier Grainger Supplier No. 14L752
The Telemecanique XCSLF-14L752 is a safety interlock switch with a metal head and plastic body. It has a 14mm actuator with a maximum switching capacity of 6A at 250V AC. This interlock switch has gold plated contacts and is IP67 rated for protection against dust and temporary immersion. It is designed for use on safety guards and access panels to prevent operation when open. This interlock switch provides reliable position detection for industrial safety applications.