Learn More
Grainger Safety Interlock Switch Zamak 2NO/1NC
Catalog No. 502751662
Encompass_Preferred
The Allen-Bradley 802T-N4A1 rectangular magnetic safety interlock switch by Rockwell Automation is a non-contact interlock switch that uses a magnetic field for position sensing. It has a compact rectangular plastic housing suitable for guard door applications. The operating range is up to 10mm and it is rated for safety category 4 per ISO 13849-1. This interlock switch has 2 NO and 1 NC contacts with a maximum switching capacity of 6A/250VAC. It can help improve machine safety by disabling dangerous machine motion when a guard door is opened. With its robust design, this interlock switch provides reliable and cost-effective protection for industrial equipment.
Catalog No. 50-275-1662 Supplier Grainger Supplier No. 14L748
The Allen-Bradley 802T-N4A1 rectangular magnetic safety interlock switch by Rockwell Automation is a non-contact interlock switch that uses a magnetic field for position sensing. It has a compact rectangular plastic housing suitable for guard door applications. The operating range is up to 10mm and it is rated for safety category 4 per ISO 13849-1. This interlock switch has 2 NO and 1 NC contacts with a maximum switching capacity of 6A/250VAC. It can help improve machine safety by disabling dangerous machine motion when a guard door is opened. With its robust design, this interlock switch provides reliable and cost-effective protection for industrial equipment.