Learn More
Grainger Safety Interlock Switch Zamak 3NC
Catalog No. 502751676
Encompass_Preferred
The Telemecanique XCSLF-14L753 is a safety interlock switch with a metal head and plastic body. It has a 14mm actuator with a maximum switching capacity of 6A at 250VAC. This interlock switch is designed for use in safety applications to guard machine access points. It features direct opening operation, snap action contacts, and gold plated double break contacts. The switch body offers IP67 protection.
Catalog No. 50-275-1676 Supplier Grainger Supplier No. 14L753
The Telemecanique XCSLF-14L753 is a safety interlock switch with a metal head and plastic body. It has a 14mm actuator with a maximum switching capacity of 6A at 250VAC. This interlock switch is designed for use in safety applications to guard machine access points. It features direct opening operation, snap action contacts, and gold plated double break contacts. The switch body offers IP67 protection.