Learn More
SCIMAC Safety Lab Stopper, Extraction Nut, Standard Taper, Hollow, PTFE, Joint: 19/38, Hollow
Catalog No. NC3239443
Encompass_Preferred
Safety Lab Stopper, Extraction Nut, Standard Taper, Hollow, PTFE, Joint: 19/38, Hollow, Height: 55mm.  Full Length; Safety Lab Solid PTFE| Black Extraction Nut | Easy Removal from glass Joints | Boiling & Distilling Flasks |Autoclave Safe at 250˚F | Complies to ASTM E675
Catalog No. NC3239443 Supplier SCIMAC Supplier No. SM83113
Safety Lab Stopper, Extraction Nut, Standard Taper, Hollow, PTFE, Joint: 19/38, Hollow, Height: 55mm.  Full Length; Safety Lab Solid PTFE| Black Extraction Nut | Easy Removal from glass Joints | Boiling & Distilling Flasks |Autoclave Safe at 250˚F | Complies to ASTM E675