Learn More
Accuform™ Safety Label, CAUTION AREA IN FRONT OF THIS ELECTRICAL
Catalog No. 19111141 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Vinyl
Catalog No. Material
19-111-141 Vinyl
1 options
This item is not returnable. View return policy
Catalog No. 19-111-141 Supplier Accuform™ Supplier No. LELC601VSP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Labels identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Printed matte surface has permanent acrylic adhesive back
  • Flexible vinyl can be applied to curved surfaces
  • Moisture resistant for a variety of light-duty indoor and sheltered outdoor marking needs
  • Square corners and scored back liner for quick peel-off and application
  • 3.5 x 5, Adhesive Vinyl, Pk. 5
Specifications
Material Vinyl
Quantity 5 Pk.
Size (English) 3.5 x 5 in.
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Displayed Text CAUTION AREA IN FRONT OF ELECTRICAL PANEL MUST BE KEPT CLEAR
Color Black on Yellow