Learn More
Accuform™ Safety Label, WARNING ARC FLASH AND SHOCK HAZARD
Catalog No. 19111143 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Vinyl
Catalog No. Material
19-111-143 Vinyl
1 options
Catalog No. 19-111-143 Supplier Accuform™ Supplier No. LELC309
Add to Cart
Add to Cart
Labels identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
  • Printed image protected by UV protective coating for durability indoor and outdoor marking locations
  • Permanent acrylic adhesive back
  • Dual-layer flexibility can be applied to curved areas
  • Round corners and scored back liner for quick peel-off and application
  • 3 x 5, adhesive dura-vinyl
Specifications
Material Vinyl
Size (English) 3.5 x 5 in.
Mounting Type Self-adhesive
Shape Rectangular
Languages English
Displayed Text WARNING ARC FLASH AND SHOCK HAZARD
Color Yellow on White