Learn More
Fisherbrand™ Safety Multi-Sample Needles
Catalog No. 22024234 Shop All Fisherbrand Products
Change view
Click to view available options
Gauge:
21 G
22 G
Length (English):
1.25 in.
Preattached Holder:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Length (English) Preattached Holder
22-024-234 22 G 1.25 in. Yes
22-024-233 21 G 1.25 in. No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-024-234
  • PRODUCT PROMOTION
    $109.56
    $78.75 / Pack of 50
    Save $30.81
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • PRODUCT PROMOTION
    $795.07
    $630.00 / Case of 8 PK
    Save $165.07
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fisherbrand Safety Multi-Sample Needles make it possible to collect several samples with a single vein puncture.
  • Low-angle bevel design allows easy penetration
  • Color-coded caps and hubs allow easy identification of needle gauge size
  • Available with safety mechanism covering the needle after use to guard against accidental needlestick during normal handling
Specifications
Sterility Sterile
Disposable Single-use
Gauge 22 G
Latex Free Yes
Length (English) 1.25 in.
Preattached Holder Yes