Learn More
Med Vet International BD Safety Hypodermic Needle Eclipse™, 1 Inch Length, 23 Gauge, Thin Wall, Hinged Safety Needle, 100/Box
Catalog No. 503043408
Encompass_Preferred
Features Pivoting Shielding Mechanism No hard surface required for activation Bevel--oriented needle allows for low-angle injections BD PrecisionGlide™ needles minimize injection pain and patient discomfort Needle-based design facilitates standardization and SKU reduction For use with BD Luer-Lok™ syringes only
Catalog No. 50-304-3408 Supplier Med Vet International Supplier No. 305762
Features Pivoting Shielding Mechanism No hard surface required for activation Bevel--oriented needle allows for low-angle injections BD PrecisionGlide™ needles minimize injection pain and patient discomfort Needle-based design facilitates standardization and SKU reduction For use with BD Luer-Lok™ syringes only