Learn More
Med Vet International BD Safety Hypodermic Needle Eclipse™, 1/2 Inch Length, 27 Gauge, Thin Wall, Hinged Safety Needle, 100/Box
Catalog No. 503043510
Encompass_Preferred
Features Single-handed activation. No hard surface required for activation
Catalog No. 50-304-3510 Supplier Med Vet International Supplier No. 305758
Features Single-handed activation. No hard surface required for activation