Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Safety Pendant Cover Large

Catalog No. 19129734 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-734 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-734 Supplier Brady™ Supplier No. 151252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Have a mechanical hoist or overhead crane that’s not in use? Keep it that way by controlling access to the buttons with the Brady Pendant Control Safety Cover.

  • Expandable design fits 12 to 39 inch long pendants
  • Prevent accidental access with rigid sides that protect buttons
  • Store it away – collapsible design allows it to flatten so you can easily store it when not in use
  • Keep the cover secured on the pendant control by applying up to four padlocks

Specifications

Product Type Switch and Fuse Protection
Color Red
Material Canvas
Description Expandable design fits 12 to 39 in. long pendants
For Use With (Equipment) Pendant Control
Quantity 1 Ea.

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.